Ethyl 4-(methylamino)-2-(methylsulfanyl)-5-pyrimidinecarboxylate 97%

Supplier: Ambeed
Warning

A466017-1G A466017-250MG A466017-5G
77034-246EA 49.87 USD
77034-246 77034-248 77034-250
Ethyl 4-(methylamino)-2-(methylsulfanyl)-5-pyrimidinecarboxylate 97%
Ethyl 4-(methylamino)-2-(methylsulfanyl)-5-pyrimidinecarboxylate

Precaución: for Research Use Only

Formula: C₉H₁₃N₃O₂S
MW: 227.28 g/mol
MDL Number: MFCD00476788
CAS Number: 76360-82-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR