3-Amino-6-chloro-4,5-dimethylpyridazine 98%

Supplier: Ambeed
Warning

A465674-250MG A465674-5G A465674-1G
77004-450EA 12 USD
77004-450 77004-452 77882-183
3-Amino-6-chloro-4,5-dimethylpyridazine 98%
3-Amino-6-chloro-4,5-dimethylpyridazine

Precaución: for Research Use Only

Formula: C₆H₈ClN₃
MDL Number: MFCD00828815
CAS Number: 76593-36-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR