2-Chlorothieno[3,2-d]pyrimidine 97%

Supplier: Ambeed
Warning

A465485-250MG A465485-25G A465485-5G A465485-1G
77034-232EA 23 USD
77034-232 77034-234 77034-236 77602-034
2-Chlorothieno[3,2-d]pyrimidine 97%
2-Chlorothieno[3,2-d]pyrimidine

Precaución: for Research Use Only

Formula: C₆H₃ClN₂S
MDL Number: MFCD12406127
CAS Number: 1119280-68-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR