2-(3-Methylisoxazol-5-yl)acetic acid 98%

Supplier: Ambeed
Warning

A464976-1G A464976-250MG A464976-5G A464976-100G A464976-25G
76990-862EA 11 USD
76990-862 76990-864 76990-866 77890-944 77890-945
2-(3-Methylisoxazol-5-yl)acetic acid 98%
2-(3-Methylisoxazol-5-yl)acetic acid

Precaución: for Research Use Only

Formula: C₆H₇NO₃
MDL Number: MFCD01863539
CAS Number: 19668-85-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR