8-Amino-3-bromoimidazo[1,2-a]pyrazine 98%

Supplier: Ambeed
Warning

A464565-100MG A464565-250MG A464565-5G
77016-884EA 24 USD
77016-884 77016-886 77016-888
8-Amino-3-bromoimidazo[1,2-a]pyrazine 98%
8-Amino-3-bromoimidazo[1,2-a]pyrazine

Precaución: for Research Use Only

Formula: C₆H₅BrN₄
MDL Number: MFCD09265736
CAS Number: 117718-92-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR