(1,3-Bis(2,4,6-trimethylphenyl)-2-imidazolidinylidene)dichloro(phenylmethylene)(tricyclohexylphosphine)ruthenium 97%

Supplier: Ambeed
Danger

A464512-10G A464512-1G A464512-5G A464512-25G
77034-226EA 320.4 USD
77034-226 77034-228 77034-230 77572-464
(1,3-Bis(2,4,6-trimethylphenyl)-2-imidazolidinylidene)dichloro(phenylmethylene)(tricyclohexylphosphine)ruthenium 97%
(1,3-Bis(2,4,6-trimethylphenyl)-2-imidazolidinylidene)dichloro(phenylmethylene)(tricyclohexylphosphine)ruthenium

Precaución: for Research Use Only

Formula: C₄₆H₆₅Cl₂N₂PRu
MW: 848.97 g/mol
MDL Number: MFCD03453237
CAS Number: 246047-72-3
UN: 1325
ADR: 4.1,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR