2-Bromo-9,10-diphenylanthracene 98%

Supplier: Ambeed
Warning

A464076-100G A464076-1G A464076-25G A464076-5G
76990-852EA 415.88 USD
76990-852 76990-854 76990-856 76990-858
2-Bromo-9,10-diphenylanthracene 98%
2-Bromo-9,10-diphenylanthracene

Precaución: for Research Use Only

Formula: C₂₆H₁₇Br
MW: 409.33 g/mol
MDL Number: MFCD14708167
CAS Number: 201731-79-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR