4,4'-Diaminobiphenyl-2,2'-disulfonic acid hydrate 70%

Supplier: Ambeed
Warning

Synonyms: 2,2'-Benzidinedisulphonic Acid, 4,4'-Diamino-[1,1'-biphenyl]-2,2'-disulphonic acid(contains 30% Water at maximum)

A463990-1000G A463990-100G A463990-25G
77004-432EA 66.84 USD
77004-432 77004-434 77004-436
4,4'-Diaminobiphenyl-2,2'-disulfonic acid hydrate 70%
4,4'-Diaminobiphenyl-2,2'-disulfonic acid hydrate

Precaución: for Research Use Only

Formula: C₁₂H₁₂N₂O₆S₂·xH₂O
MW: 344.36 g/mol (anhy)
MDL Number: MFCD00041885
CAS Number: 117-61-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR