(R)-3-(Methylamino)-1-(thiophen-2-yl)propan-1-ol 95%

Supplier: Ambeed
Warning

A463778-250MG A463778-1G
76990-850EA 171 USD
76990-850 77886-209
(R)-3-(Methylamino)-1-(thiophen-2-yl)propan-1-ol 95%
(R)-3-(Methylamino)-1-(thiophen-2-yl)propan-1-ol

Precaución: for Research Use Only

Formula: C₈H₁₃NOS
MDL Number: MFCD09026399
CAS Number: 116539-57-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR