(5-chloro-6-fluoropyridin-3-yl)boronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 3-Chloro-2-fluoropyridne-5-boronic acid, (5-Chloro-6-fluoropyridin-3-yl)boronic acid

A463639-100MG A463639-1G A463639-250MG
77016-870EA 37.14 USD
77016-870 77016-872 77016-874
(5-chloro-6-fluoropyridin-3-yl)boronic acid 98%
(5-chloro-6-fluoropyridin-3-yl)boronic acid

Precaución: for Research Use Only

Formula: C₅H₄BClFNO₂
MW: 175.353 g/mol
MDL Number: MFCD18257900
CAS Number: 1366482-32-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR