2,4-Dibromo-5-fluoropyridine 95%

Supplier: Ambeed
Warning

A463441-100MG A463441-1G A463441-250MG A463441-5G
76990-842EA 45.62 USD
76990-842 76990-844 76990-846 76990-848
2,4-Dibromo-5-fluoropyridine 95%
2,4-Dibromo-5-fluoropyridine

Precaución: for Research Use Only

Formula: C₅H₂Br₂FN
MW: 254.88 g/mol
MDL Number: MFCD11114541
CAS Number: 1256805-06-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR