3-Bromo-5-(1,3,4-oxadiazol-2-yl)pyridine 95%

Supplier: Ambeed
Warning

Synonyms: 2-(5-Bromopyridin-3-yl)-1,3,4-oxadiazole

A462995-100MG A462995-1G A462995-250MG
77004-422EA 11 USD
77004-422 77004-424 77004-426
3-Bromo-5-(1,3,4-oxadiazol-2-yl)pyridine 95%
3-Bromo-5-(1,3,4-oxadiazol-2-yl)pyridine

Precaución: for Research Use Only

Formula: C₇H₄BrN₃O
MW: 226.04 g/mol
MDL Number: MFCD08444031
CAS Number: 924869-13-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR