5-Chloro-3-fluoro-2-iodopyridine 95%

Supplier: Ambeed
Warning

A462966-1G A462966-250MG
76990-838EA 320.4 USD
76990-838 76990-840
5-Chloro-3-fluoro-2-iodopyridine 95%
5-Chloro-3-fluoro-2-iodopyridine

Precaución: for Research Use Only

Formula: C₅H₂ClFIN
MW: 257.43 g/mol
MDL Number: MFCD18256751
CAS Number: 514797-98-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR