1-Chloro-2-(4-ethoxybenzyl)-4-iodobenzene 98%
Supplier: Ambeed
|
Warning
|
A462402-100G
A462402-10G
A462402-1G
A462402-250MG
A462402-25G
A462402-5G
76990-826EA
280.08
USD
76990-826
76990-828
76990-830
76990-832
76990-834
76990-836
1-Chloro-2-(4-ethoxybenzyl)-4-iodobenzene 98%
1-Chloro-2-(4-ethoxybenzyl)-4-iodobenzene
Precaución: for Research Use Only
Formula:
C₁₅H₁₄ClIO MW: 372.62 g/mol |
MDL Number:
MFCD18642361 CAS Number: 1103738-29-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...