5-Amino-3-(4-chlorophenyl)-1-phenylpyrazole 98%

Supplier: Ambeed
Warning

Synonyms: 3-(4-Chlorophenyl)-1-phenyl-1H-pyrazol-5-amine

A462284-1G A462284-250MG A462284-5G
77016-858EA 105.03 USD
77016-858 77016-860 77016-862
5-Amino-3-(4-chlorophenyl)-1-phenylpyrazole 98%
5-Amino-3-(4-chlorophenyl)-1-phenylpyrazole

Precaución: for Research Use Only

Formula: C₁₅H₁₂ClN₃
MW: 269.73 g/mol
MDL Number: MFCD00197059
CAS Number: 19652-14-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR