tert-Butyl 4-carbamoylbenzylcarbamate 95%

Supplier: Ambeed
Warning

A461827-100MG A461827-1G
77016-854EA 23 USD
77016-854 77016-856
tert-Butyl 4-carbamoylbenzylcarbamate 95%
tert-Butyl 4-carbamoylbenzylcarbamate

Precaución: for Research Use Only

Formula: C₁₃H₁₈N₂O₃
MDL Number: MFCD24389093
CAS Number: 871721-44-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR