1-Benzhydrylazetidine-3-carboxylic acid 98%

Supplier: Ambeed
Warning

Synonyms: 1-(Diphenylmethyl)azetidine-3-carboxylic Acid

A461796-1G A461796-25G A461796-5G
77034-198EA 10.61 USD
77034-198 77034-200 77034-202
1-Benzhydrylazetidine-3-carboxylic acid 98%
1-Benzhydrylazetidine-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₇H₁₇NO₂
MW: 267.33 g/mol
MDL Number: MFCD01566538
CAS Number: 36476-87-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR