1-(Mercaptomethyl)cyclopropaneacetic acid 95%

Supplier: Ambeed
Danger

Synonyms: [1-(Thiomethyl)cycloprop-1-yl]acetic acid, 2-(1-(Mercaptomethyl)cyclopropyl)acetic acid

A461701-1G A461701-25G A461701-5G
77004-416EA 12.74 USD
77004-416 77004-418 77004-420
1-(Mercaptomethyl)cyclopropaneacetic acid 95%
1-(Mercaptomethyl)cyclopropaneacetic acid

Precaución: for Research Use Only

Formula: C₆H₁₀O₂S
MW: 146.20 g/mol
MDL Number: MFCD03411600
CAS Number: 162515-68-6
UN: 3261
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR