Benzyl (1-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)cyclopropyl)carbamate 95%

Supplier: Ambeed
Warning

A460785-100MG A460785-1G A460785-250MG
77016-840EA 39 USD
77016-840 77016-842 77016-844
Benzyl (1-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)cyclopropyl)carbamate 95%
Benzyl (1-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)cyclopropyl)carbamate

Precaución: for Research Use Only

Formula: C₂₃H₂₈BNO₄
MW: 393.28 g/mol
MDL Number: MFCD16618967
CAS Number: 1256359-24-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR