7-Hydroxy-4-methyl-2-oxo-2H-chromene-8-carbaldehyde 98%

Supplier: Ambeed
Warning

Synonyms: 4μ8C

A460722-10MG A460722-1MG A460722-25MG A460722-50MG A460722-5MG A460722-250MG
77004-406EA 57 USD
77004-406 77004-408 77004-410 77004-412 77004-414 77673-174
7-Hydroxy-4-methyl-2-oxo-2H-chromene-8-carbaldehyde 98%
7-Hydroxy-4-methyl-2-oxo-2H-chromene-8-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₁H₈O₄
MDL Number: MFCD12027255
CAS Number: 14003-96-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR