3-(3-Fluorophenoxy)propionic acid 95%

Supplier: Ambeed
Warning

A460613-1G A460613-250MG
77004-404EA 43 USD
77004-404 77673-172
3-(3-Fluorophenoxy)propionic acid 95%
3-(3-Fluorophenoxy)propionic acid

Precaución: for Research Use Only

Formula: C₉H₉FO₃
MDL Number: MFCD01931913
CAS Number: 133077-42-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR