Palladium(II) trimethylacetate 98%

Supplier: Ambeed
Warning

Synonyms: Palladium(II) pivalate

A460236-1G A460236-250MG A460236-100MG A460236-5G
76990-814EA 185.66 USD
76990-814 76990-816 77613-422 77684-250
Palladium(II) trimethylacetate 98%
Palladium(II) trimethylacetate

Precaución: For research use only.

Formula: C₁₀H₁₈O₄Pd
MW: 308.67 g/mol
MDL Number: MFCD16038138
CAS Number: 106224-36-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR