4-Bromo-3-fluorobenzamide 96%

Supplier: Ambeed
Warning

A460191-1G A460191-250MG A460191-5G
77016-830EA 35 USD
77016-830 77016-832 77016-834
4-Bromo-3-fluorobenzamide 96%
4-Bromo-3-fluorobenzamide

Precaución: for Research Use Only

Formula: C₇H₅BrFNO
MDL Number: MFCD06409121
CAS Number: 759427-20-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR