1-Phenylvinylboronic acid pinacol ester 95%, Ambeed.Inc

Supplier: Ambeed
Warning

Synonyms: 4,4,5,5-Tetramethyl-2-(1-phenylvinyl)-1,3,2-dioxaborolane

A460019-25G A460019-5G A460019-1G
76990-810EA 142 USD
76990-810 76990-812 77684-249
1-Phenylvinylboronic acid pinacol ester 95%, Ambeed.Inc
1-Phenylvinylboronic acid pinacol ester

Precaución: For research use only.

Formula: C₁₄H₁₉BO₂
MW: 230.11 g/mol
MDL Number: MFCD06659923
CAS Number: 143825-84-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR