(1,2-Dihydroacenaphthylen-5-yl)boronic acid 96%

Supplier: Ambeed
Warning

A460018-1G A460018-250MG A460018-5G
77016-826EA 36 USD
77016-826 77016-828 77890-933
(1,2-Dihydroacenaphthylen-5-yl)boronic acid 96%
(1,2-Dihydroacenaphthylen-5-yl)boronic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₁BO₂
MDL Number: MFCD00448048
CAS Number: 183158-33-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR