tert-Butyl 4-(6-aminopyrimidin-4-yl)piperazine-1-carboxylate 95%

Supplier: Ambeed
Warning

A459742-1G A459742-250MG
77016-822EA 192.03 USD
77016-822 77016-824
tert-Butyl 4-(6-aminopyrimidin-4-yl)piperazine-1-carboxylate 95%
tert-Butyl 4-(6-aminopyrimidin-4-yl)piperazine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₃H₂₁N₅O₂
MW: 279.33 g/mol
MDL Number: MFCD11109805
CAS Number: 436851-80-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR