Ethyl-trans-3-(1-pyrrolidinyl)acrylate 95%

Supplier: Ambeed
Warning

A459279-25G A459279-5G A459279-1G
77034-186EA 397 USD
77034-186 77034-188 77872-759
Ethyl-trans-3-(1-pyrrolidinyl)acrylate 95%
Ethyl-trans-3-(1-pyrrolidinyl)acrylate

Precaución: for Research Use Only

Formula: C₉H₁₅NO₂
MDL Number: MFCD01075735
CAS Number: 65651-80-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR