(R)-3-Amino-4-(2,4,5-trifluorophenyl)butyric acid 95%

Supplier: Ambeed
Warning

Synonyms: (R)-3-Amino-4-(2,4,5-trifluorophenyl)butanoic acid

A459217-10G A459217-1G A459217-25G
77034-180EA 250.38 USD
77034-180 77034-182 77034-184
(R)-3-Amino-4-(2,4,5-trifluorophenyl)butyric acid 95%
(R)-3-Amino-4-(2,4,5-trifluorophenyl)butyric acid

Precaución: for Research Use Only

Formula: C₁₀H₁₀F₃NO₂
MW: 233.187 g/mol
MDL Number: MFCD07363507
CAS Number: 936630-57-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR