2-Fluoro-1H-imidazole hydrochloride 95%, Ambeed.Inc

Supplier: Ambeed
Warning

A458839-100MG A458839-250MG
77016-820EA 191 USD
77016-820 77684-247
2-Fluoro-1H-imidazole hydrochloride 95%, Ambeed.Inc
2-Fluoro-1H-imidazole hydrochloride

Precaución: For research use only.

Formula: C₃H₄ClFN₂
MW: 122.53 g/mol
MDL Number: MFCD24369564
CAS Number: 57212-34-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR