3,5-Dichloro-4-fluoroaniline 98%

Supplier: Ambeed
Warning

A458733-10G A458733-1G A458733-250MG
77034-174EA 25.47 USD
77034-174 77034-176 77034-178
3,5-Dichloro-4-fluoroaniline 98%
3,5-Dichloro-4-fluoroaniline

Precaución: for Research Use Only

Formula: C₆H₄Cl₂FN
MW: 180.007 g/mol
MDL Number: MFCD09835492
CAS Number: 2729-34-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR