Ethyl 4-amino-5-bromonicotinate 98%

Supplier: Ambeed
Warning

A458707-250MG A458707-5G
77016-818EA 64.72 USD
77016-818 77684-246
Ethyl 4-amino-5-bromonicotinate 98%
Ethyl 4-amino-5-bromonicotinate

Precaución: For research use only.

Formula: C₈H₉BrN₂O₂
MW: 245.07 g/mol
MDL Number: MFCD16988618
CAS Number: 1240595-43-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR