5-Bromo-2-(cyclopropylamino)pyrimidine 98%

Supplier: Ambeed
Warning

A458572-100MG A458572-1G A458572-250MG A458572-5G
76990-802EA 5 USD
76990-802 76990-804 76990-806 76990-808
5-Bromo-2-(cyclopropylamino)pyrimidine 98%
5-Bromo-2-(cyclopropylamino)pyrimidine

Precaución: for Research Use Only

Formula: C₇H₈BrN₃
MDL Number: MFCD03646032
CAS Number: 886366-20-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR