Fmoc-Glu-OFm 97%

Supplier: Ambeed
Warning

Synonyms: (S)-5-((9H-Fluoren-9-yl)methoxy)-4-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-5-oxopentanoic acid

A458197-1G A458197-250MG
76990-798EA 62.6 USD
76990-798 76990-800
Fmoc-Glu-OFm 97%
Fmoc-Glu-OFm

Precaución: for Research Use Only

Formula: C₃₄H₂₉NO₆
MW: 547.607 g/mol
MDL Number: MFCD00237656
CAS Number: 200616-18-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR