tert-Butyl-4-(chloromethyl)tetrahydro-1(2H)-pyridinecarboxylate 97%

Supplier: Ambeed
Danger

Synonyms: 1-Boc-4-(chloromethyl)piperidine, 4-(Chloromethyl)piperidine, N-BOC protected

A458191-1G A458191-250MG A458191-10G A458191-25G A458191-5G
77004-398EA 41 USD
77004-398 77004-400 77882-172 77882-173 77882-174
tert-Butyl-4-(chloromethyl)tetrahydro-1(2H)-pyridinecarboxylate 97%
tert-Butyl-4-(chloromethyl)tetrahydro-1(2H)-pyridinecarboxylate

Precaución: for Research Use Only

Formula: C₁₁H₂₀ClNO₂
MDL Number: MFCD08689676
CAS Number: 479057-79-9
UN: 1759
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR