2-Methoxy-3-nitrobenzoic acid 98%

Supplier: Ambeed
Warning

A458012-1G A458012-250MG A458012-25G A458012-5G
77016-810EA 35 USD
77016-810 77016-812 77016-814 77016-816
2-Methoxy-3-nitrobenzoic acid 98%
2-Methoxy-3-nitrobenzoic acid

Precaución: for Research Use Only

Formula: C₈H₇NO₅
MDL Number: MFCD11044854
CAS Number: 40751-88-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR