5-Fluoro-2-methoxyisonicotinaldehyde 98%

Supplier: Ambeed
Warning

A457934-100MG A457934-1G A457934-250MG
77034-168EA 95.49 USD
77034-168 77034-170 77034-172
5-Fluoro-2-methoxyisonicotinaldehyde 98%
5-Fluoro-2-methoxyisonicotinaldehyde

Precaución: for Research Use Only

Formula: C₇H₆FNO₂
MW: 155.126 g/mol
MDL Number: MFCD08277260
CAS Number: 884495-12-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR