Methyl 6-chloro-4-methylnicotinate 97%

Supplier: Ambeed
Warning

Synonyms: Methyl 2-chloro-4-methylpyridine-5-carboxylate

A457046-100MG A457046-1G A457046-250MG A457046-5G
76990-790EA 37.14 USD
76990-790 76990-792 76990-794 76990-796
Methyl 6-chloro-4-methylnicotinate 97%
Methyl 6-chloro-4-methylnicotinate

Precaución: for Research Use Only

Formula: C₈H₈ClNO₂
MW: 185.60 g/mol
MDL Number: MFCD18258050
CAS Number: 1224464-97-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR