2-((3-Oxo-3-phenylpropyl)amino)acetic acid 98%

Supplier: Ambeed
Warning

Synonyms: N-(3-PhenylPropionyl)-Gly-OH

A456802-1G A456802-250MG A456802-100MG
76990-786EA 144.29 USD
76990-786 76990-788 77684-245
2-((3-Oxo-3-phenylpropyl)amino)acetic acid 98%
2-((3-Oxo-3-phenylpropyl)amino)acetic acid

Precaución: For research use only.

Formula: C₁₁H₁₃NO₃
MW: 207.22 g/mol
MDL Number: MFCD00749241
CAS Number: 56613-60-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR