2-Bromo-6-methoxybenzamide 95%

Supplier: Ambeed
Warning

A456748-100MG A456748-250MG
77016-806EA 41.38 USD
77016-806 77016-808
2-Bromo-6-methoxybenzamide 95%
2-Bromo-6-methoxybenzamide

Precaución: for Research Use Only

Formula: C₈H₈BrNO₂
MW: 230.06 g/mol
MDL Number: MFCD18391869
CAS Number: 1261581-85-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR