Dibenzosuberone 98%

Supplier: Ambeed
Warning

Synonyms: 1-Dibenzo[b,f]cycloheptanone, 10,11-Dihydro-5H-dibenzo[a,d][7]annulen-5-one, 5-Dibenzosuberone, Dibenzo[b,f]cycloheptan-1-one

A456403-1G A456403-25G A456403-5G A456403-500G A456403-100G
77004-368EA 5.31 USD
77004-368 77004-370 77004-372 77568-900 77601-970
Dibenzosuberone 98%
Dibenzosuberone

Precaución: For research use only.

Formula: C₁₅H₁₂O
MW: 208.26 g/mol
MDL Number: MFCD00003587
CAS Number: 1210-35-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR