(1,3-Dioxo-1,3-dihydro-2H-isoindol-2-yl)acetonitrile ≥98%
Supplier: Ambeed
|
Warning
|
A456176-1G
A456176-250MG
A456176-25G
A456176-5G
77004-360EA
16
USD
77004-360
77004-362
77004-364
77004-366
(1,3-Dioxo-1,3-dihydro-2H-isoindol-2-yl)acetonitrile ≥98%
(1,3-Dioxo-1,3-dihydro-2H-isoindol-2-yl)acetonitrile
Precaución: for Research Use Only
Formula:
C₁₀H₆N₂O₂ |
MDL Number:
MFCD00087362 CAS Number: 3842-20-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...