Methyl 6-cyano-1-naphthoate 95+%

Supplier: Ambeed
Warning

A456116-100MG A456116-1G A456116-250MG
77016-800EA 39 USD
77016-800 77016-802 77016-804
Methyl 6-cyano-1-naphthoate 95+%
Methyl 6-cyano-1-naphthoate

Precaución: for Research Use Only

Formula: C₁₃H₉NO₂
MDL Number: MFCD27997156
CAS Number: 91804-22-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR