(2R,3R,4R,5S)-6-(Methylamino)hexane-1,2,3,4,5-pentaol ((3-(((2R,3S)-2-((R)-1-(3,5-bis(trifluoromethyl)phenyl)ethoxy)-3-(4-fluorophenyl)morpholino)methyl)-5-oxo-4,5-dihydro-1H-1,2,4-triazol-1-yl)phosphonate)(2:1) 98%

Supplier: Ambeed
Warning

Synonyms: Fosaprepitant Dimeglumine

A455841-100MG A455841-10MG A455841-1MG A455841-50MG A455841-5MG
77034-156EA 29.71 USD
77034-156 77034-158 77034-160 77034-162 77034-164
(2R,3R,4R,5S)-6-(Methylamino)hexane-1,2,3,4,5-pentaol ((3-(((2R,3S)-2-((R)-1-(3,5-bis(trifluoromethyl)phenyl)ethoxy)-3-(4-fluorophenyl)morpholino)methyl)-5-oxo-4,5-dihydro-1H-1,2,4-triazol-1-yl)phosphonate)(2:1) 98%
(2R,3R,4R,5S)-6-(Methylamino)hexane-1,2,3,4,5-pentaol ((3-(((2R,3S)-2-((R)-1-(3,5-bis(trifluoromethyl)phenyl)ethoxy)-3-(4-fluorophenyl)morpholino)methyl)-5-oxo-4,5-dihydro-1H-1,2,4-triazol-1-yl)phosphonate)(2:1)

Precaución: for Research Use Only

Formula: C₃₇H₅₆F₇N₆O₁₆P
MW: 1004.83 g/mol
MDL Number: MFCD23102059
CAS Number: 265121-04-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR