3-(4'-Fluorobenzyloxy)phenylboronic acid 95%

Supplier: Ambeed
Warning

Synonyms: (3-((4-Fluorobenzyl)oxy)phenyl)boronic acid

A455777-100MG A455777-1G A455777-250MG A455777-5G
77016-792EA 6 USD
77016-792 77016-794 77016-796 77016-798
3-(4'-Fluorobenzyloxy)phenylboronic acid 95%
3-(4'-Fluorobenzyloxy)phenylboronic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₂BFO₃
MDL Number: MFCD08705238
CAS Number: 1072952-03-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR