5-Cyclopropyl-2-methoxynicotinaldehyde 97%

Supplier: Ambeed
Warning

A455641-100MG A455641-1G A455641-250MG
76990-778EA 53 USD
76990-778 76990-780 76990-782
5-Cyclopropyl-2-methoxynicotinaldehyde 97%
5-Cyclopropyl-2-methoxynicotinaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₁₁NO₂
MDL Number: MFCD19690244
CAS Number: 888499-98-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR