6,6-Diphenylfulvene 98%

Supplier: Ambeed
Warning

Synonyms: 5-(Diphenylmethylene)-1,3-cyclopentadiene

A455560-1G A455560-250MG A455560-25G A455560-5G
77034-148EA 42.44 USD
77034-148 77034-150 77034-152 77034-154
6,6-Diphenylfulvene 98%
6,6-Diphenylfulvene

Precaución: for Research Use Only

Formula: C₁₈H₁₄
MW: 230.31 g/mol
MDL Number: MFCD00001419
CAS Number: 2175-90-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR