(S)-(+)-2-Methylpiperazine 97%

Supplier: Ambeed
Danger

A455418-100G A455418-25G A455418-5G
76990-772EA 71 USD
76990-772 76990-774 76990-776
(S)-(+)-2-Methylpiperazine 97%
(S)-(+)-2-Methylpiperazine

Precaución: for Research Use Only

Formula: C₅H₁₂N₂
MDL Number: MFCD00171452
CAS Number: 74879-18-8
UN: 1325
ADR: 4.1,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR