Glutaconaldehydedianil hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: N-(5-Anilino-2,4-pentadienylidene)aniline hydrochloride, N-[5-(Phenylamino)-2,4-pentadienylidene]aniline monohydrochloride, N-((2E,4E)-5-(Phenylamino)penta-2,4-dien-1-ylidene)aniline hydrochloride

A454132-100G A454132-10G A454132-25G A454132-500G A454132-5G
77004-342EA 118.83 USD
77004-342 77004-344 77004-346 77004-348 77004-350
Glutaconaldehydedianil hydrochloride 97%
Glutaconaldehydedianil hydrochloride

Precaución: for Research Use Only

Formula: C₁₇H₁₆N₂
MW: 284.79 g/mol
MDL Number: MFCD00012630
CAS Number: 1497-49-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR