3-Chloro-5-(trifluoromethoxy)aniline 95%

Supplier: Ambeed
Danger

A453675-100MG A453675-1G A453675-250MG
77034-126EA 15.92 USD
77034-126 77034-128 77034-130
3-Chloro-5-(trifluoromethoxy)aniline 95%
3-Chloro-5-(trifluoromethoxy)aniline

Precaución: for Research Use Only

Formula: C₇H₅ClF₃NO
MW: 211.57 g/mol
MDL Number: MFCD09832245
CAS Number: 151276-13-0
UN: 2810
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR