Fmoc-Val-Cit-OH 95%

Supplier: Ambeed
Warning

Synonyms: (S)-2-((S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-methylbutanamido)-5-ureidopentanoic acid

A453269-250MG A453269-25G A453269-5G A453269-1G
76990-766EA 11.67 USD
76990-766 76990-768 76990-770 77594-420
Fmoc-Val-Cit-OH 95%
Fmoc-Val-Cit-OH

Precaución: for Research Use Only

Formula: C₂₆H₃₂N₄O₆
MW: 496.555 g/mol
MDL Number: MFCD28139060
CAS Number: 159858-21-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR